Ethyl 7-chloro-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110440-1G
APOSOR110440-1GEA 149 EUR
APOSOR110440-1G
Ethyl 7-chloro-1-benzothiophene-2-carboxylate
Ethyl 7-chloro-1-benzothiophene-2-carboxylate
Formel: C₁₁H₉ClO₂S
Molecular Weight: 240,706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00834770
CAS-Nummer: 90407-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR