8-Chloroquinazolin-2-amine

Lieferant: Apollo Scientific
Achtung

OR110441-1G
APOSOR110441-1GEA 232 EUR
APOSOR110441-1G
8-Chloroquinazolin-2-amine
8-Chloroquinazolin-2-amine
Formel: C₈H₆ClN₃
Molecular Weight: 179,606 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565604
CAS-Nummer: 1185113-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR