(5-Methoxy-1H-indol-3-yl)methanol

Lieferant: Apollo Scientific
Achtung

OR110443-1G
APOSOR110443-1GEA 232 EUR
APOSOR110443-1G
(5-Methoxy-1H-indol-3-yl)methanol
(5-Methoxy-1H-indol-3-yl)methanol
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202621
CAS-Nummer: 77419-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR