Methyl 2-chloro-6-(homopiperazin-1-yl)isonicotinate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-chloro-6-(1,4-diazepan-1-yl)isonicotinate

OR110445-1G
APOSOR110445-1GEA 0 EUR
APOSOR110445-1G
Methyl 2-chloro-6-(homopiperazin-1-yl)isonicotinate
Methyl 2-chloro-6-(homopiperazin-1-yl)isonicotinate
Formel: C₁₂H₁₆ClN₃O₂
Molecular Weight: 269,727 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037414
CAS-Nummer: 1427460-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR