Methyl 1-methyl-1H-indole-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Methoxycarbonyl)-1-methyl-1H-indole
OR110447-1G
APOSOR110447-1GEA
87
EUR
APOSOR110447-1G
Methyl 1-methyl-1H-indole-4-carboxylate
Methyl 1-methyl-1H-indole-4-carboxylate
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,211 g/mol Schmelzpunkt: 49…51 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09839048 CAS-Nummer: 1444-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...