Methyl 1-methyl-1H-indole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110449-1G
APOSOR110449-1GEA 75.4 EUR
APOSOR110449-1G
Methyl 1-methyl-1H-indole-3-carboxylate
Methyl 1-methyl-1H-indole-3-carboxylate
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,211 g/mol
Schmelzpunkt: 82…84 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00085570
CAS-Nummer: 108438-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR