4-Pentylbenzamide

Lieferant: Apollo Scientific

OR110451-1G OR110451-250MG OR110451-5G
APOSOR110451-1GEA 186 EUR
APOSOR110451-1G APOSOR110451-250MG APOSOR110451-5G
4-Pentylbenzamide
4-Pentylbenzamide
Formel: C₁₂H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00984217
CAS-Nummer: 138913-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR