N-Ethyl-4-pentylbenzamide

Lieferant: Apollo Scientific

OR110453-1G OR110453-250MG OR110453-5G
APOSOR110453-1GEA 0 EUR
APOSOR110453-1G APOSOR110453-250MG APOSOR110453-5G
N-Ethyl-4-pentylbenzamide
N-Ethyl-4-pentylbenzamide
Formel: C₁₄H₂₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03551084
CAS-Nummer: 401587-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR