(5-Methoxy-1-methyl-1H-indol-3-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR110460-1G
APOSOR110460-1GEA
476
EUR
APOSOR110460-1G
(5-Methoxy-1-methyl-1H-indol-3-yl)methanol
(5-Methoxy-1-methyl-1H-indol-3-yl)methanol
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191,226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00983073 CAS-Nummer: 313535-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...