2-(4-Formylphenoxy)nicotinonitrile

Lieferant: Apollo Scientific

OR110471-1G
APOSOR110471-1GEA 248 EUR
APOSOR110471-1G
2-(4-Formylphenoxy)nicotinonitrile
2-(4-Formylphenoxy)nicotinonitrile
Formel: C₁₃H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08741856
CAS-Nummer: 919731-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR