6-(4-Formylphenoxy)nicotinonitrile

Lieferant: Apollo Scientific

OR110474-1G
APOSOR110474-1GEA 248 EUR
APOSOR110474-1G
6-(4-Formylphenoxy)nicotinonitrile
6-(4-Formylphenoxy)nicotinonitrile
Formel: C₁₃H₈N₂O₂
Molecular Weight: 224,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756245
CAS-Nummer: 676501-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR