4-(Benzyloxy)-5-methoxy-2-nitrobenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR110477-1G
APOSOR110477-1GEA
125
EUR
APOSOR110477-1G
4-(Benzyloxy)-5-methoxy-2-nitrobenzaldehyd
4-(Benzyloxy)-5-methoxy-2-nitrobenzaldehyd
Formel:
C₁₅H₁₃NO₅ Molecular Weight: 287,2674 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02629357 CAS-Nummer: 2426-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...