1-(3-Methyl-4-nitrophenyl)pyrrolidine

Lieferant: Apollo Scientific

OR110479-1G
APOSOR110479-1GEA 173 EUR
APOSOR110479-1G
1-(3-Methyl-4-nitrophenyl)pyrrolidine
1-(3-Methyl-4-nitrophenyl)pyrrolidine
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11037903
CAS-Nummer: 218139-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR