1-Methyl-4-(3-methyl-4-nitrophenyl)piperazine

Lieferant: Apollo Scientific

OR110482-1G
APOSOR110482-1GEA 0 EUR
APOSOR110482-1G
1-Methyl-4-(3-methyl-4-nitrophenyl)piperazine
1-Methyl-4-(3-methyl-4-nitrophenyl)piperazine
Formel: C₁₂H₁₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11039716
CAS-Nummer: 16154-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR