1-(3-Methyl-4-nitrophenyl)-1H-imidazole
Lieferant: Apollo Scientific
OR110483-1G
APOSOR110483-1GEA
296
EUR
APOSOR110483-1G
1-(3-Methyl-4-nitrophenyl)-1H-imidazole
1-(3-Methyl-4-nitrophenyl)-1H-imidazole
Formel:
C₁₀H₉N₃O₂ Molecular Weight: 203,197 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11870465 CAS-Nummer: 102791-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...