5-Bromo-2,4-dichlorobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR110485-1G
APOSOR110485-1GEA 122 EUR
APOSOR110485-1G
5-Bromo-2,4-dichlorobenzoic acid
5-Bromo-2,4-dichlorobenzoic acid
Formel: C₇H₃BrCl₂O₂
Schmelzpunkt: 181…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09910550
CAS-Nummer: 791137-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR