1-Phenylcyclobutanecarboxamide

Lieferant: Apollo Scientific

OR110488-1G
APOSOR110488-1GEA 236 EUR
APOSOR110488-1G
1-Phenylcyclobutanecarboxamide
1-Phenylcyclobutanecarboxamide
Formel: C₁₁H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD10664451
CAS-Nummer: 17380-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR