[2-(4-Bromophenoxy)phenyl]methanol

Lieferant: Apollo Scientific

OR110491-1G
APOSOR110491-1GEA 248 EUR
APOSOR110491-1G
[2-(4-Bromophenoxy)phenyl]methanol
[2-(4-Bromophenoxy)phenyl]methanol
Formel: C₁₃H₁₁BrO₂
Molecular Weight: 279,129 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11190400
CAS-Nummer: 74744-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR