[4-(4-Bromophenoxy)phenyl]methanol

Lieferant: Apollo Scientific

OR110492-1G
APOSOR110492-1GEA 248 EUR
APOSOR110492-1G
[4-(4-Bromophenoxy)phenyl]methanol
[4-(4-Bromophenoxy)phenyl]methanol
Formel: C₁₃H₁₁BrO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11190398
CAS-Nummer: 1039899-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR