1-Bromo-4-[4-(bromomethyl)phenoxy]benzene

Lieferant: Apollo Scientific

OR110494-1G
APOSOR110494-1GEA 310 EUR
APOSOR110494-1G
1-Bromo-4-[4-(bromomethyl)phenoxy]benzene
1-Bromo-4-[4-(bromomethyl)phenoxy]benzene
Formel: C₁₃H₁₀Br₂O
Molecular Weight: 342,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146806
CAS-Nummer: 1344045-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR