2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde

Lieferant: Apollo Scientific
Achtung

OR110496-1G OR110496-5G
APOSOR110496-1GEA 125 EUR
APOSOR110496-1G APOSOR110496-5G
2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde
2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde
Formel: C₁₇H₁₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01568764
CAS-Nummer: 181297-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR