1-(Bromomethyl)-2-(4-tert-butylphenoxy)benzene

Lieferant: Apollo Scientific

OR110510-1G
APOSOR110510-1GEA 322 EUR
APOSOR110510-1G
1-(Bromomethyl)-2-(4-tert-butylphenoxy)benzene
1-(Bromomethyl)-2-(4-tert-butylphenoxy)benzene
Formel: C₁₇H₁₉BrO
Molecular Weight: 319,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619591
CAS-Nummer: 1427461-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR