1-(Bromomethyl)-2-(3-tert-butylphenoxy)benzene

Lieferant: Apollo Scientific

OR110515-1G
APOSOR110515-1GEA 322 EUR
APOSOR110515-1G
1-(Bromomethyl)-2-(3-tert-butylphenoxy)benzene
1-(Bromomethyl)-2-(3-tert-butylphenoxy)benzene
Formel: C₁₇H₁₉BrO
Molecular Weight: 319,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619595
CAS-Nummer: 1427460-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR