1-(Bromomethyl)-4-(4-tert-butylphenoxy)benzene

Lieferant: Apollo Scientific

OR110528-1G
APOSOR110528-1GEA 409 EUR
APOSOR110528-1G
1-(Bromomethyl)-4-(4-tert-butylphenoxy)benzene
1-(Bromomethyl)-4-(4-tert-butylphenoxy)benzene
Formel: C₁₇H₁₉BrO
Molecular Weight: 319,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619599
CAS-Nummer: 1427461-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR