4'-(Bromomethyl)-2-chlorobiphenyl

Lieferant: Apollo Scientific

Synonyme: 4-(2-Chlorophenyl)benzyl bromide, 4'-(Bromomethyl)-2-chloro-1,1'-biphenyl

OR110534-1G
APOSOR110534-1GEA 433 EUR
APOSOR110534-1G
4'-(Bromomethyl)-2-chlorobiphenyl
4'-(Bromomethyl)-2-chlorobiphenyl
Formel: C₁₃H₁₀BrCl
Molecular Weight: 281,576 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146809
CAS-Nummer: 210113-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR