4-(4-Bromophenoxy)-2-methylaniline

Lieferant: Apollo Scientific

OR110536-1G
APOSOR110536-1GEA 248 EUR
APOSOR110536-1G
4-(4-Bromophenoxy)-2-methylaniline
4-(4-Bromophenoxy)-2-methylaniline
Formel: C₁₃H₁₂BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD12169249
CAS-Nummer: 1154154-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR