2-Methyl-4-phenoxyaniline
Lieferant: Apollo Scientific
OR110537-1G
APOSOR110537-1GEA
149
EUR
APOSOR110537-1G
2-Methyl-4-phenoxyaniline
2-Methyl-4-phenoxyaniline
Formel:
C₁₃H₁₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08688317 CAS-Nummer: 13024-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...