2-Methyl-4-(1-pyrrolidinyl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
OR110538-1G
APOSOR110538-1GEA
0
EUR
APOSOR110538-1G
2-Methyl-4-(1-pyrrolidinyl)aniline
2-Methyl-4-(1-pyrrolidinyl)aniline
Formel:
C₁₁H₁₆N₂ Molecular Weight: 176,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06337021 CAS-Nummer: 143525-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...