2-Methyl-4-(4-morpholinyl)aniline

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-4-(4-morpholinyl)benzenamine

OR110540-1G
APOSOR110540-1GEA 0 EUR
APOSOR110540-1G
2-Methyl-4-(4-morpholinyl)aniline
2-Methyl-4-(4-morpholinyl)aniline
Formel: C₁₁H₁₆N₂O
Molecular Weight: 192,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10686817
CAS-Nummer: 581-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR