5-Bromo-2-(4-methylphenoxy)pyridine

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-(p-tolyloxy)pyridine

OR110554-1G
APOSOR110554-1GEA 310 EUR
APOSOR110554-1G
5-Bromo-2-(4-methylphenoxy)pyridine
5-Bromo-2-(4-methylphenoxy)pyridine
Formel: C₁₂H₁₀BrNO
Molecular Weight: 264,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14652098
CAS-Nummer: 1248915-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR