4-(2-Oxopyrrolidin-1-yl)benzenesulphonamide

Lieferant: Apollo Scientific

OR110557-1G
APOSOR110557-1GEA 221 EUR
APOSOR110557-1G
4-(2-Oxopyrrolidin-1-yl)benzenesulphonamide
4-(2-Oxopyrrolidin-1-yl)benzenesulphonamide
Formel: C₁₀H₁₂N₂O₃S
Molecular Weight: 240,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00453080
CAS-Nummer: 36090-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR