tert-Butyl 4-{2-[(2-chloropyridin-3-yl)oxy]ethyl}piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR110563-1G
APOSOR110563-1GEA 296 EUR
APOSOR110563-1G
tert-Butyl 4-{2-[(2-chloropyridin-3-yl)oxy]ethyl}piperazine-1-carboxylate
tert-Butyl 4-{2-[(2-chloropyridin-3-yl)oxy]ethyl}piperazine-1-carboxylate
Formel: C₁₆H₂₄ClN₃O₃
Molecular Weight: 341.833 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111411
CAS-Nummer: 1227954-32-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR