3-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone

Lieferant: Apollo Scientific

OR110566-1G
APOSOR110566-1GEA 198 EUR
APOSOR110566-1G
3-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
3-[2-(4-tert-Butoxycarbonylpiperazin-1-yl)ethoxy]acetophenone
Formel: C₁₉H₂₈N₂O₄
Molecular Weight: 348,437 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109538
CAS-Nummer: 351076-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR