4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazole

OR110575-1G
APOSOR110575-1GEA 114 EUR
APOSOR110575-1G
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole
Formel: C₈H₁₁BrN₂O
Molecular Weight: 231.1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505042
CAS-Nummer: 1040377-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR