Methyl [5-(2-amino-1,3-thiazol-4-yl)-1-methyl-1H-pyrrol-2-yl]acetate

Lieferant: Apollo Scientific

OR110578-1G
APOSOR110578-1GEA 371 EUR
APOSOR110578-1G
Methyl [5-(2-amino-1,3-thiazol-4-yl)-1-methyl-1H-pyrrol-2-yl]acetate
Methyl [5-(2-amino-1,3-thiazol-4-yl)-1-methyl-1H-pyrrol-2-yl]acetate
Formel: C₁₁H₁₃N₃O₂S
Molecular Weight: 251,305 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11973598
CAS-Nummer: 1227954-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR