(3-Methoxythien-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

OR110579-1G
APOSOR110579-1GEA 0 EUR
APOSOR110579-1G
(3-Methoxythien-2-yl)methanol
(3-Methoxythien-2-yl)methanol
Formel: C₆H₈O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD11846258
CAS-Nummer: 220139-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR