tert-Butyl 4-[2-(2-formylphenoxy)ethyl]piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR110582-1G
APOSOR110582-1GEA 248 EUR
APOSOR110582-1G
tert-Butyl 4-[2-(2-formylphenoxy)ethyl]piperazine-1-carboxylate
tert-Butyl 4-[2-(2-formylphenoxy)ethyl]piperazine-1-carboxylate
Formel: C₁₈H₂₆N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD11973599
CAS-Nummer: 1227954-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR