tert-Butyl 4-[2-(2-bromophenoxy)ethyl]piperazine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110585-1G
APOSOR110585-1GEA 248 EUR
APOSOR110585-1G
tert-Butyl 4-[2-(2-bromophenoxy)ethyl]piperazine-1-carboxylate
tert-Butyl 4-[2-(2-bromophenoxy)ethyl]piperazine-1-carboxylate
Formel: C₁₇H₂₅BrN₂O₃
Molecular Weight: 385,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11973602
CAS-Nummer: 1227954-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR