2-(1H-Pyrazol-1-yl)isonicotinonitrile

Lieferant: Apollo Scientific

OR110591-1G
APOSOR110591-1GEA 0 EUR
APOSOR110591-1G
2-(1H-Pyrazol-1-yl)isonicotinonitrile
2-(1H-Pyrazol-1-yl)isonicotinonitrile
Formel: C₉H₆N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09373409
CAS-Nummer: 956769-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR