4-(4-Bromo-1H-pyrazol-1-yl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

OR110600-1G
APOSOR110600-1GEA 371 EUR
APOSOR110600-1G
4-(4-Bromo-1H-pyrazol-1-yl)benzenesulphonyl chloride
4-(4-Bromo-1H-pyrazol-1-yl)benzenesulphonyl chloride
Formel: C₉H₆BrClN₂O₂S
Molecular Weight: 321.573 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406835
CAS-Nummer: 1174064-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR