2-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde

Lieferant: Apollo Scientific

OR110606-1G
APOSOR110606-1GEA 223 EUR
APOSOR110606-1G
2-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde
2-(4-Bromo-1H-pyrazol-1-yl)benzaldehyde
Formel: C₁₀H₇BrN₂O
Molecular Weight: 251,079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756233
CAS-Nummer: 1174064-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR