tert-Butyl 4-(3-methoxy-3-oxopropyl)homopiperazine-1-carboxylate

Lieferant: Apollo Scientific

OR110608-1G
APOSOR110608-1GEA 248 EUR
APOSOR110608-1G
tert-Butyl 4-(3-methoxy-3-oxopropyl)homopiperazine-1-carboxylate
tert-Butyl 4-(3-methoxy-3-oxopropyl)homopiperazine-1-carboxylate
Formel: C₁₄H₂₆N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD05664953
CAS-Nummer: 874801-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR