5-[(Dimethylamino)sulphonyl]-2-methylbenzoic acid

Lieferant: Apollo Scientific

OR110610-1G
APOSOR110610-1GEA 173 EUR
APOSOR110610-1G
5-[(Dimethylamino)sulphonyl]-2-methylbenzoic acid
5-[(Dimethylamino)sulphonyl]-2-methylbenzoic acid
Formel: C₁₀H₁₃NO₄S
Molecular Weight: 243.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06353543
CAS-Nummer: 89001-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR