Methyl 6-chloro-2-oxo-2H-chromene-3-carboxylate

Lieferant: Apollo Scientific

OR110613-1G
APOSOR110613-1GEA 87 EUR
APOSOR110613-1G
Methyl 6-chloro-2-oxo-2H-chromene-3-carboxylate
Methyl 6-chloro-2-oxo-2H-chromene-3-carboxylate
Formel: C₁₁H₇ClO₄
Molecular Weight: 238,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04406156
CAS-Nummer: 91058-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR