5-tert-Butyl-3-(1H-pyrrol-1-yl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR110618-1G
APOSOR110618-1GEA 0 EUR
APOSOR110618-1G
5-tert-Butyl-3-(1H-pyrrol-1-yl)thiophene-2-carboxylic acid
5-tert-Butyl-3-(1H-pyrrol-1-yl)thiophene-2-carboxylic acid
Formel: C₁₃H₁₅NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD13191326
CAS-Nummer: 1305711-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR