tert-Butyl 4-hydroxy-4-[(methylamino)methyl]piperidine-1-carboxylate

Lieferant: Apollo Scientific

OR110619-1G
APOSOR110619-1GEA 296 EUR
APOSOR110619-1G
tert-Butyl 4-hydroxy-4-[(methylamino)methyl]piperidine-1-carboxylate
tert-Butyl 4-hydroxy-4-[(methylamino)methyl]piperidine-1-carboxylate
Formel: C₁₂H₂₄N₂O₃
Molecular Weight: 244,331 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11920519
CAS-Nummer: 1073559-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR