1-Thieno[2,3-d]pyrimidin-4-ylpiperidin-4-one

Lieferant: Apollo Scientific

OR110623-1G
APOSOR110623-1GEA 371 EUR
APOSOR110623-1G
1-Thieno[2,3-d]pyrimidin-4-ylpiperidin-4-one
1-Thieno[2,3-d]pyrimidin-4-ylpiperidin-4-one
Formel: C₁₁H₁₁N₃OS
Molecular Weight: 233,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11176817
CAS-Nummer: 1094427-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR