3-Bromothiophene-2-carbohydrazide

Lieferant: Apollo Scientific

OR110624-1G
APOSOR110624-1GEA 279 EUR
APOSOR110624-1G
3-Bromothiophene-2-carbohydrazide
3-Bromothiophene-2-carbohydrazide
Formel: C₅H₅BrN₂OS
Molecular Weight: 221,075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036203
CAS-Nummer: 1043593-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR