4-(8-methylimidazo[1,2-a]pyridin-2-yl)aniline

Lieferant: Apollo Scientific
Achtung

OR110626-1G OR110626-5G OR110626-10G
APOSOR110626-1GEA 173 EUR
APOSOR110626-1G APOSOR110626-5G APOSOR110626-10G
4-(8-methylimidazo[1,2-a]pyridin-2-yl)aniline
4-(8-methylimidazo[1,2-a]pyridin-2-yl)aniline
Formel: C₁₄H₁₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04971851
CAS-Nummer: 365565-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR