4-(2-Oxo-1-pyrrolidinyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

OR110629-1G OR110629-5G
APOSOR110629-1GEA 270 EUR
APOSOR110629-1G APOSOR110629-5G
4-(2-Oxo-1-pyrrolidinyl)benzenesulphonyl chloride
4-(2-Oxo-1-pyrrolidinyl)benzenesulphonyl chloride
Formel: C₁₀H₁₀ClNO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00219329
CAS-Nummer: 112539-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR